Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038112.3 | NP_001033201.1 | 4 - 5 | 471 | 117 | |
GenBank mRNA | BC109932.1 | - | 4 - 5 | 513 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |