Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBATA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038121.2 | NP_001033210.2 | 2 - 3 | 150 | 64 | |
GenBank mRNA | BC110031.1 | - | 2 - 3 | 151 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |