Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SQOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040511.2 | NP_001035601.2 | 2 - 3 | 278 | 66 | |
GenBank mRNA | BC104605.1 | - | 2 - 3 | 278 | 66 | |
BT021504.1 | - | 2 - 3 | 344 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |