Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046094.2 | NP_001039559.2 | 1 - 2 | 78 | 77 | |
GenBank mRNA | EE386336.1 | - | 1 - 2 | 78 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |