Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046094.2 | NP_001039559.2 | 3 - 4 | 346 | 110 | |
GenBank mRNA | BC112782.1 | - | 2 - 3 | 268 | 110 | |
EE386336.1 | - | 3 - 4 | 346 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |