Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP5KL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075847.2 | NP_001069315.2 | 1 - 2 | 93 | 156 | |
GenBank mRNA | BC118210.1 | - | 1 - 2 | 93 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |