Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V0E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097574.2 | NP_001091043.1 | 1 - 2 | 176 | 59 | |
GenBank mRNA | BC112699.1 | - | 1 - 2 | 177 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |