Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097997.1 | NP_001091466.1 | 3 - 4 | 379 | 84 | |
GenBank mRNA | BC114657.1 | - | 3 - 4 | 379 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |