Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097997.1 | NP_001091466.1 | 4 - 5 | 1803 | 68 | |
GenBank mRNA | BC114657.1 | - | 4 - 5 | 1803 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |