Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPZL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098051.1 | NP_001091520.1 | 5 - 6 | 716 | 129 | |
GenBank mRNA | BC140504.1 | - | 5 - 6 | 716 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |