Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098054.1 | NP_001091523.1 | 3 - 4 | 690 | 112 | |
GenBank mRNA | BC140598.1 | - | 3 - 4 | 690 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |