Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098058.1 | NP_001091527.1 | 9 - 10 | 1897 | 127 | |
GenBank mRNA | BC149569.1 | - | 9 - 10 | 2201 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |