Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BFAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098869.1 | NP_001092339.1 | 6 - 7 | 1219 | 88 | |
GenBank mRNA | BC142184.1 | - | 6 - 7 | 1219 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |