Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098886.1 | NP_001092356.1 | 343 | 119 | ||
GenBank mRNA | BC142105.1 | - | 343 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus |