Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098895.2 | NP_001092365.1 | 4 - 5 | 444 | 78 | |
GenBank mRNA | BC146143.1 | - | 4 - 5 | 445 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |