Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098904.1 | NP_001092374.1 | 1 - 2 | 120 | 85 | |
GenBank mRNA | BC146183.1 | - | 1 - 2 | 120 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |