Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098904.1 | NP_001092374.1 | 9 - 10 | 1085 | 87 | |
GenBank mRNA | BC146183.1 | - | 9 - 10 | 1085 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |