Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098909.2 | NP_001092379.1 | 7 - 8 | 930 | 87 | |
GenBank mRNA | BC142205.1 | - | 6 - 7 | 1199 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |