Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098918.2 | NP_001092388.1 | 3 - 4 | 274 | 97 | |
GenBank mRNA | BC146158.1 | - | 3 - 4 | 275 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |