Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098918.2 | NP_001092388.1 | 4 - 5 | 466 | 77 | |
GenBank mRNA | BC146158.1 | - | 4 - 5 | 467 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |