Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098933.1 | NP_001092403.1 | 2 - 3 | 444 | 61 | |
GenBank mRNA | BC146216.1 | - | 2 - 3 | 444 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |