Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098940.1 | NP_001092410.1 | 8 - 9 | 954 | 107 | |
GenBank mRNA | BC142122.1 | - | 8 - 9 | 954 | 107 | |
BC149564.1 | - | 8 - 9 | 969 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |