Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098953.1 | NP_001092423.1 | 1 - 2 | 245 | 103 | |
GenBank mRNA | BC142157.1 | - | 1 - 2 | 245 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |