Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098968.2 | NP_001092438.1 | 4 - 5 | 597 | 58 | |
GenBank mRNA | BC142260.1 | - | 4 - 5 | 598 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |