Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098982.2 | NP_001092452.1 | 6 - 7 | 3167 | 66 | |
GenBank mRNA | BC142415.1 | - | 6 - 7 | 3168 | 66 | |
BC151288.1 | - | 6 - 7 | 2915 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |