Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098982.2 | NP_001092452.1 | 7 - 8 | 3245 | 86 | |
GenBank mRNA | BC142415.1 | - | 7 - 8 | 3246 | 86 | |
BC151288.1 | - | 7 - 8 | 2993 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |