Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098988.2 | NP_001092458.1 | 7 - 8 | 785 | 78 | |
GenBank mRNA | BC146269.1 | - | 7 - 8 | 787 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |