Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TULP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098992.1 | NP_001092462.1 | 4 - 5 | 487 | 64 | |
GenBank mRNA | BC142186.1 | - | 4 - 5 | 487 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |