Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099001.1 | NP_001092471.1 | 7 - 8 | 837 | 68 | |
GenBank mRNA | BC142478.1 | - | 7 - 8 | 837 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |