Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099010.2 | NP_001092480.1 | 5 - 6 | 814 | 102 | |
GenBank mRNA | BC142528.1 | - | 5 - 6 | 815 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |