Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099015.2 | NP_001092485.1 | 5 - 6 | 1124 | 87 | |
GenBank mRNA | BC146268.1 | - | 5 - 6 | 1125 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |