Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099015.2 | NP_001092485.1 | 9 - 10 | 1644 | 77 | |
GenBank mRNA | BC146268.1 | - | 9 - 10 | 1645 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |