Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIPK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100327.1 | NP_001093797.1 | 3 - 4 | 623 | 76 | |
GenBank mRNA | BC148094.1 | - | 3 - 4 | 623 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |