Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIPK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100327.1 | NP_001093797.1 | 6 - 7 | 941 | 61 | |
GenBank mRNA | BC148094.1 | - | 6 - 7 | 941 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |