Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMAA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100342.2 | NP_001093812.1 | 5 - 6 | 1034 | 112 | |
GenBank mRNA | BC149854.1 | - | 5 - 6 | 1030 | 112 | |
BC149875.1 | - | 5 - 6 | 1038 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |