Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100351.1 | NP_001093821.1 | 2 - 3 | 232 | 76 | |
GenBank mRNA | BC149445.1 | - | 2 - 3 | 232 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |