Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100363.2 | NP_001093833.1 | 6 - 7 | 942 | 100 | |
GenBank mRNA | BC149966.1 | - | 6 - 7 | 943 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |