Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100376.1 | NP_001093846.1 | 5 - 6 | 1441 | 66 | |
GenBank mRNA | BC114036.1 | - | 6 - 7 | 1582 | 66 | |
BC150086.1 | - | 5 - 6 | 1441 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |