Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RESF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101072.2 | NP_001094542.1 | 6 - 7 | 5437 | 128 | |
GenBank mRNA | BC151612.1 | - | 6 - 7 | 5438 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |