Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PACSIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101101.1 | NP_001094571.1 | 5 - 6 | 806 | 65 | |
GenBank mRNA | BC151572.1 | - | 5 - 6 | 806 | 65 | |
BC153241.1 | - | 4 - 5 | 675 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |