Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGAT2L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101861.1 | NP_001095331.2 | 5 - 6 | 712 | 101 | |
GenBank mRNA | BC149181.1 | - | 5 - 6 | 712 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |