Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGAT2L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101861.1 | NP_001095331.2 | 6 - 7 | 919 | 76 | |
GenBank mRNA | BC149181.1 | - | 6 - 7 | 919 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |