Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101913.2 | NP_001095383.1 | 5 - 6 | 989 | 68 | |
GenBank mRNA | BC150047.1 | - | 5 - 6 | 990 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |