Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH3A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101984.1 | NP_001095454.1 | 8 - 9 | 1181 | 106 | |
GenBank mRNA | BC149984.1 | - | 8 - 9 | 1181 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |