Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102038.1 | NP_001095508.1 | 10 - 11 | 1163 | 73 | |
GenBank mRNA | BC148039.1 | - | 10 - 11 | 1163 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |