Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KANK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102053.2 | NP_001095523.2 | 4 - 5 | 2374 | 77 | |
GenBank mRNA | BC149084.1 | - | 6 - 7 | 887 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |