Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102063.1 | NP_001095533.1 | 2 - 3 | 471 | 61 | |
GenBank mRNA | BC151573.1 | - | 2 - 3 | 471 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |