Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102077.1 | NP_001095547.1 | 8 - 9 | 1321 | 102 | |
GenBank mRNA | BC149049.1 | - | 8 - 9 | 1321 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |