Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1B10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102088.2 | NP_001095558.1 | 1 - 2 | 161 | 72 | |
GenBank mRNA | BC151501.1 | - | 1 - 2 | 162 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |