Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102378.1 | NP_001095848.1 | 6 - 7 | 1262 | 99 | |
GenBank mRNA | BC149862.1 | - | 6 - 7 | 1262 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |